Learn More
Product Code | Brand | Description | For Use With (Equipment) | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Equipment) | Includes | Price | Quantity & Availability | ||||
10608515
|
Thermo Scientific™
290MA201 |
290 MA201 | Thermo Scientific TRACE 1300/1310 SSL and ISQ GC-MS Interface Kits | Pk. of 10 Ferrules |
932.00 NOK
Pack of 10 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
||||
10670616
|
Thermo Scientific™
290MA202 |
290 MA202 | All SilTite Kits | Pk. of 10 Ferrules |
971.00 NOK
Pack of 10 |
Estimated Shipment: 30-06-2025
Log in to see stock. |
||||
10659275
|
Thermo Scientific™
290MA203 |
290 MA203 | All SilTite Kits | Pk. of 10 Ferrules |
971.00 NOK
Pack of 10 |
Estimated Shipment: 30-06-2025
Log in to see stock. |
||||
10724077
|
Thermo Scientific™
290MA205 |
290 MA205 | MS Interface | Pk. of 5 Nuts |
1155.00 NOK
Pack of 10 |
In Stock
Log in to see stock. |
||||
10391145
|
Thermo Scientific™
290MA207 |
290 MA207 | Agilent SSL Injectors | Pk. of 5 Nuts |
1605.00 NOK
Pack of 5 |
Estimated Shipment: 30-06-2025
Log in to see stock. |
||||
10565625
|
Thermo Scientific™
290MT211 |
290 MT211 | Thermo Scientific DSQ GC-MS Interface | Pk. of 5 Nuts |
1415.00 NOK
Pack of 10 |
Estimated Shipment: 07-07-2025
Log in to see stock. |
||||
10698695
|
Thermo Scientific™
290MT221 |
290 MT221 | Thermo Scientific DSQ GC-MS Interface Kits | Pk. of 10 Ferrules |
961.00 NOK
Pack of 10 |
Estimated Shipment: 30-06-2025
Log in to see stock. |
||||
10713677
|
Thermo Scientific™
290MT222 |
290 MT222 | Thermo Scientific DSQ GC-MS Interface Kits | Pk. of 10 Ferrules |
971.00 NOK
Pack of 10 |
Estimated Shipment: 04-07-2025
Log in to see stock. |
||||
10321395
|
Thermo Scientific™
290MT223 |
290 MT223 | Thermo Scientific DSQ GC-MS Interface Kits | Pk. of 10 Ferrules |
971.00 NOK
Pack of 10 |
Estimated Shipment: 09-07-2025
Log in to see stock. |
||||
Description
Eliminate leaks with the Thermo Scientific™ SilTite Ferrule, which forms a strong, permanent, airtight seal around the capillary column.
- SilTite metal ferrules and nuts are made from the same material and have the same thermal
- expansion coefficient
- Base of the ferrule is flat and forms a perfect seal with the MS interface
- The ferrule's temperature tolerance is well above the limit of the injector, MS interface or GC oven
- Unlike other ferrules, do not need re-tightening after installation