Learn More
BRAND™ SilberBrand™ Burette Tube for Compact Burette
Borosilicate glass construction. Replacement burette tube for compact burette.
Brand:  BRAND™ 13938
258.54 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10404732
  • 328.00 NOK / Each
Only null left
Estimated Shipment: 21-07-2025
Description
  • SilberBrand™ borosilicate glass (3.3) construction
  • Replacement burette tube
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 mL
Compact Burette
Borosilicate Glass
Buret Tube
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BRAND™ SilberBrand™ Burette Tube for Compact Burette > Capacity: 50 mL