Learn More
BRAND™ SilberBrand™ Burette with Lateral Stopcock
Boro 3.3 glass class B burette with lateral stopcock
Brand:  BRAND™ 13568
1979.43 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15211329
  • 2515.00 NOK / Pack of 2
Only null left
Estimated Shipment: 19-08-2025
Description
  • Boro 3.3 Glass
  • To DIN EN ISO 385
  • Calibrated to deliver (TD, Ex)
  • Black Schellbach stripe
  • Interchangeable needle-valve and glass key
  • Length approx. 800 mm
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    50 mL volume with 0.1 mL subdivisions
    Borosilicate Glass (3.3)
    2
    Documents
    One moment while we fetch your results.
    Certificates
    Promotions
    Promotions
    Product Content Correction
    Product Title
    BRAND™ SilberBrand™ Burette with Lateral Stopcock > Description: 50 mL volume with 0.1 mL subdivisions