missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIL1 Monoclonal Antibody (OTI3E3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500997

This item is not returnable.
View return policy
Description
This gene encodes a resident endoplasmic reticulum (ER), N-linked glycoprotein with an N-terminal ER targeting sequence, 2 putative N-glycosylation sites, and a C-terminal ER retention signal. This protein functions as a nucleotide exchange factor for another unfolded protein response protein. Mutations in this gene have been associated with Marinesco-Sjogren syndrome. Alternate transcriptional splice variants have been characterized.Specifications
SIL1 | |
Monoclonal | |
0.56 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9H173 | |
SIL1 | |
Full length human recombit protein of human SIL1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3E3 | |
Unconjugated | |
SIL1 | |
BAP, MSS, ULG5 | |
Mouse | |
Affinity Chromatography | |
RUO | |
64374 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 12
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
SIL1 Monoclonal Antibody (OTI3E3), TrueMAB™, OriGene
Spot an opportunity for improvement?Share a Content Correction