missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIK1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1705.02 NOK - 4821.09 NOK
Specifications
Antigen | SIK1 |
---|---|
Concentration | 133 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16806325
|
Proteintech
51045-1-AP-20UL |
20 μL |
1705.02 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16896315
|
Proteintech
51045-1-AP-150UL |
150 μL |
4821.09 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SIK1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SIK1. This antibody recognizes human, mouse antigen. The SIK1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
SIK1 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Human, Mouse | |
IgG |
133 μg/mL | |
Polyclonal | |
Rabbit | |
150094 | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title