Learn More
Epredia™ Signature Series™ Clarifier™ 1, 2
Enhance the nuclear definition and clarity of Signature Series hematoxylins with Epredia™ Signature Series Clarifier™ 1, 2.
Brand: Epredia™ 7401
351.96 NOK valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
- Improves contrast between Nuclei and Cytoplasm
- Increases cell transparency
- Removes background staining caused by excessive tissue adhesive
- Removes unwanted hematoxylin staining from adhesive without affecting nuclear staining
- Clarifier 1 is recommended with Hematoxylin 7211 and Hematoxylin 1
- Clarifier 2 is recommended with Hematoxylin 7212 and Hematoxylin 2
Specifications
Clarifier 1 | |
1 gal. |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H371-May cause damage to organs.
- EUH210-Safety data sheet available on request.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P321-Specific treatment (see on this label).
- P370+P378-In case of fire: Use to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.