Learn More
Siglec-2/CD22 Antibody (RFB4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-49890
Product Code. 18610527
  • 4505.00 NOK / 0.20mg
Description
Siglec-2/CD22 Monoclonal antibody specifically detects Siglec-2/CD22 in Human, Canine samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Siglec-2/CD22
Monoclonal
Unconjugated
B-cell receptor CD22, BL-CAM, B-lymphocyte cell adhesion molecule, CD22 antigenMGC130020, CD22 molecule, sialic acid binding Ig-like lectin 2, Sialic acid-binding Ig-like lectin 2, SIGLEC-2, SIGLEC2FLJ22814, T-cell surface antigen Leu-14
135 kDa type I integral membrane protein
0.2 mg
Adaptive Immunity, B Cell Development and Differentiation Markers, DNA Repair, Immunology
933
Store at 4°C. Do not freeze.
IgG1
Flow Cytometry, Immunofluorescence, Immunofluorescence
RFB4
PBS (pH 7.2) and 1.0% BSA
Mouse
Protein A or G purified
RUO
Primary
Human, Canine
Purified
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Siglec-2/CD22 Antibody (RFB4), Novus Biologicals™ > 0.2 mg, Unconjugated