missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ Shimadzu Split / Splitless Liner with Single Taper
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Brand: Trajan Scientific and Medical™ 092071
This item is not returnable.
View return policy
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with Shimadzu instruments
Specifications
5/Pk. | |
Split / Splitless Inlet Liner with Single Taper | |
5 mm |
95 mm | |
3.4 mm | |
Shimadzu GC Instruments |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Trajan Scientific and Medical™ Shimadzu Split / Splitless Liner with Single Taper >
Spot an opportunity for improvement?Share a Content Correction