Learn More
Fisherbrand™ Shell Vial Kit, PP, Clear, PE plug
Recommended for HPLC usage.
198.00 NOK - 493.00 NOK
Specifications
Color Clear
Material Polypropylene
No. per Pack 100
Marking Spot No
View More Specs
Products 3
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
11561404
View Documents
Fisherbrand™
08 34 2194
1.28 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12970941
View Documents
Fisherbrand™
15 34 2197
5.5 mL
230.00 NOK
Pack of 100
Only null left
Estimated Shipment: 01-07-2025
Add to basket
12980941
View Documents
Fisherbrand™
15 34 2199
4 mL
493.00 NOK
Pack of 100
Only null left
In Stock
Edge
Add to basket
Description
Description
  • Shell vials and the appropriate plugs can also be supplied as a 2in1 KIT
  • Handy and inexpensive vial/ closure combination for uncritical analyses
Specifications
Specifications
Clear
Polypropylene
100
No
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ Shell Vial Kit, PP, Clear, PE plug