Learn More
SGE™ Gastight Syringes: Fixed Needle Models
Suitable for liquid and gas samples
692.00 NOK - 3590.00 NOK
Specifications
Accuracy | ±1% |
---|---|
For Use With (Application) | Suitable for gases or liquids for maximum inertness |
Includes | Plunger protection, needle glued into barrel with epoxy solvent-resistant glue |
Product Type | Gastight Syringe |
Needle Point Style | Fixed Bevel |
Product Code | Brand | Volume (Metric) | Needle Gauge | Model | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Needle Gauge | Model | Price | Quantity & Availability | |||
10229250
|
SGE™
002200 |
10 μL | 26 G | 10F-GT |
692.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15298443
|
SGE™
002202 |
10 μL | 26 G | 10F-GT |
3590.00 NOK
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15208453
|
SGE™
002208 |
10 μL | 26 G | 10F-GT-7 |
769.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10167450
|
SGE™
004200 |
50 μL | 25 G | 25F-GT |
785.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10035540
|
SGE™
005200 |
100 μL | 25 G | 25F-GT |
861.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10702772
|
SGE™
006200 |
250 μL | 25 G | 250F-GT |
877.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10106930
|
SGE™
007200 |
500 μL | 25 G | 500F-GT |
861.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10605093
|
SGE™
008102 |
1 mL | 22 G | 1MDF-GT |
892.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12780710
|
SGE™
008502 |
2.5 mL | 22 G | 2.5MDF-GT |
902.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10546772
|
SGE™
003200 |
25 μL | 25 G | 25F-GT |
785.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Suitable for liquid and gas samples
- 10μL to 100mL capacities
- Constructed from borosilicate glass, with PTFE plunger tips and Kel-F™ Luer cones (where applicable)
- Needles align with zero mark on barrel to reduce dead volume, improve accuracy and precision to <±1%
- Replaceable plungers on all models
- Fixed-needle models have needle glued into barrel with epoxy solvent-resistant glue
Specifications
±1% | |
Plunger protection, needle glued into barrel with epoxy solvent-resistant glue | |
Fixed Bevel |
Suitable for gases or liquids for maximum inertness | |
Gastight Syringe |
Product Title
SGE™ Gastight Syringes: Fixed Needle Models