missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ BPX50 GC Capillary Columns

MS premium, low bleed phase designed for a range of EPA and pharmaceutical methods

9070.00 NOK - 15700.00 NOK

Specifications

Type GC Capillary Column
Max. Temperature 350°C
Polarity Mid polar
View More Specs

Products 4
Product Code Brand Length (Metric) Diameter (Metric) Inner Film Thickness Price Quantity & Availability  
Product Code Brand Length (Metric) Diameter (Metric) Inner Film Thickness Price Quantity & Availability  
12935561
View Documents
Trajan™
054761
30 m 0.32 mm 0.25 μm
10260.00 NOK
Each
Estimated Shipment: 08-01-2025
to see stock.
Add to basket
 
12985561
View Documents
Trajan™
054752
60 m 0.25 mm 0.25 μm
15700.00 NOK
Each
Estimated Shipment: 08-01-2025
to see stock.
Add to basket
 
12975561
View Documents
Trajan™
054741
30 m 0.15 mm 0.15 μm
9070.00 NOK
Each
Estimated Shipment: 08-01-2025
to see stock.
Add to basket
 
12694945
View Documents
Trajan™
054751
30 mm 0.25 mm 0.25 mm
9875.00 NOK
Each
Estimated Shipment: 08-01-2025
to see stock.
Add to basket
 
Description

Description

  • Mid-polarity column
  • Inert
  • Low bleed
  • High temperature
  • Ideal for a range of EPA methods and pharmaceutical applications
Specifications

Specifications

GC Capillary Column
350°C
Mid polar
Videos
SDS
Documents

Documents

Product Certifications