Learn More
Trajan™ eVol™ XR Syringe Starter Kit
eVol is a digitally controlled positive displacement dispensing system that can be programmed to reproducibly and accurately perform a wide variety of liquid handling procedures.
Brand: Trajan™ 2910200
This item is not returnable.
View return policy
Description
Includes:
- eVol™ XR Electronic Syringe unit
- Charger
- Stand
- 3 syringes: 5ÎĽL, 100ÎĽL and 1mL
Specifications
eVol XR Handle, Charger, Stand, 3 eVol Syringes with 5cm Bevel Tipped Needles: 5ÎĽL, 100ÎĽL, 1mL | |
eVol XR Starter Kit |
Product Content Correction
Product Title
Trajan™ eVol™ XR Syringe Starter Kit > w/Three Syringes