missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16841-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a protein which contains a several motifs including a ski homology region and a SET-binding region in addition to three nuclear localization signals. The encoded protein has been shown to bind the SET nuclear oncogene which is involved in DNA replication. Mutations in this gene are associated with Schinzel-Giedion midface retraction syndrome. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
SETBP1 | |
Polyclonal | |
Unconjugated | |
SETBP1 | |
KIAA0437, SEB, SET binding protein, SET binding protein 1, SETBP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
240427, 26040, 291423 | |
-20°C | |
Liquid |
Western Blot | |
0.51 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9Y6X0, Q9Z180 | |
SETBP1 | |
SETBP1 Fusion Protein Ag10532 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |