Learn More
BRAND™ Set of Special Pegs
Set of Special Pegs for use with a draining rack.
Brand:  BRAND™ 95111
279.02 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10401521
  • 287.00 NOK / Pack of 11
Only null left
Estimated Shipment: 06-08-2025
Description
  • For narrow-mouthed or small vessels
  • Can be used with test tubes, small volumetric flasks, and graduated cylinders
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polypropylene
11/Pk
6 mm
Drain Rack
100 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BRAND™ Set of Special Pegs > Material: Polypropylene