missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serum Amyloid A1 Antibody (CL9175), Novus Biologicals™
Mouse Monoclonal Antibody
3455.00 NOK - 5565.00 NOK
Specifications
Antigen | Serum Amyloid A1 |
---|---|
Clone | CL9175 |
Dilution | Western Blot 1 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18629184
|
Novus Biologicals
NBP3-21169-25ul |
25 μg |
3455.00 NOK
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18612315
|
Novus Biologicals
NBP3-21169-100ul |
100 μg |
5565.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Serum Amyloid A1 Monoclonal antibody specifically detects Serum Amyloid A1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Serum Amyloid A1 | |
Western Blot 1 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
MGC111216, PIG4, SAA, SAA2, serum amyloid A protein, serum amyloid A1, TP53I4, tumor protein p53 inducible protein 4 | |
This antibody was developed against a Synthetic Peptide of P0DJI8. Actual immunogen sequence is proprietary. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
CL9175 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Alzheimers Research, Biologically Active Proteins, Cancer, Neuroscience | |
PBS, pH 7.2, 40% glycerol | |
6288 | |
IgG1 | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Serum Amyloid A1 Antibody (CL9175), Novus Biologicals™