missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sertaconazole nitrate, Thermo Scientific™
880.62 NOK - 1598.99 NOK
Specifications
Chemical Name or Material | Sertaconazole nitrate |
---|---|
CAS | 99592-39-9 |
Melting Point | 156°C to 165°C |
Shipping Condition | No special requirements |
Product Line | Life science |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17710649
|
Thermo Scientific Acros
469250010 |
1 g |
1598.99 NOK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17720649
|
Thermo Scientific Acros
469252500 |
250 mg |
880.62 NOK
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifications
Sertaconazole nitrate | |
156°C to 165°C | |
Life science | |
Powder | |
≥97.5% |
99592-39-9 | |
No special requirements | |
C20 H15 Cl3 N2 O S . H N O3 | |
White to Beige |
Safety and Handling
Exclamation mark
Recommended Storage : Normal conditions
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title