Learn More
Falcon™ Serological Pipette, Triple Bagged
Corning’s triple-bagged packaged pipets are a safety and convenience revolution. This packaging protects the product from external contamination and is ideal for laboratory clean rooms, such as cell-based vaccine or drug production facilities.
1390.00 NOK - 5215.00 NOK
Specifications
Material | Plastic |
---|---|
Sterility | Sterile |
Graduations | Yes |
Packaging Type | Triple Bagged, Bulk |
Color | Transparent |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
30093039
|
Falcon™
355024 |
1 mL |
4595.00 NOK
Pack of 1000 |
Estimated Shipment: 30-10-2025
Log in to see stock. |
||||||
30093038
|
Falcon™
355025 |
2 mL |
4800.00 NOK
Pack of 1000 |
Estimated Shipment: 30-10-2025
Log in to see stock. |
||||||
30093041
|
Falcon™
355026 |
5 mL |
1390.00 NOK
Pack of 200 |
Estimated Shipment: 30-10-2025
Log in to see stock. |
||||||
30093040
|
Falcon™
355027 |
10 mL |
1410.00 NOK
Pack of 200 |
In Stock
Log in to see stock. |
||||||
30093042
|
Falcon™
355028 |
25 mL |
2635.00 NOK
Pack of 200 |
Estimated Shipment: 30-10-2025
Log in to see stock. |
||||||
30093043
|
Falcon™
355029 |
50 mL |
5215.00 NOK
Pack of 200 |
Estimated Shipment: 30-10-2025
Log in to see stock. |
||||||
Description
- Triple-bagged in two outer bags
- Manufactured and packaged with animal-free materials
- Sterility Assurance Level (SAL) of 10-6
- USP Class VI tested
- Non-pyrogenic
- Non-cytotoxic
- RNase-/DNase-free
- Human DNA-free
- Low particulate according to USP<788>
Specifications
Plastic | |
Yes | |
Transparent | |
Reusable |
Sterile | |
Triple Bagged, Bulk | |
Serological Pipet |
Unless otherwise specified, all products are for research use or general laboratory use only. Not intended for use in diagnostic or therapeutic procedures. Not for use in humans. These products are not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment. Corning Life Sciences makes no claims regarding the performance of these products for clinical or diagnostic applications.
Corning's products are not specifically designed and tested for diagnostic testing. Many Corning products, though not specific for diagnostic testing, can be used in the workflow and preparation of the test at the customers discretion. Customers may use these products to support their claims. We cannot make any claims or statements that our products are approved for diagnostic testing either directly or indirectly. The customer is responsible for any testing, validation, and/or regulatory submissions that may be required to support the safety and efficacy of their intended application.