Learn More
Fisherbrand™ Septum for 24-400 Screw Thread Cap
Most septa are laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.
Brand: Fisherbrand™ 22 02 0542
Description
- 22mm Septa
Specifications
Septa | |
22 mm | |
1000 |
Red-Orange, Transparent | |
Natural Rubber, TEF | |
1.3 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Product Content Correction
Product Title
Fisherbrand™ Septum for 24-400 Screw Thread Cap > N. Rubber/TEF red-orange/transparent,1.3 mm thickness,60° shore A