Learn More
Hamilton™ Septa, Long Life GC Autosampler
Brand:  Hamilton™ 75826
Product Code. 10615063
  • 1375.00 NOK / Pack of 25
Estimated Shipment: 27-03-2025
to see stock.
Explore more special offers
Specifications
Septum
400°C
25/Pk.
Documents
One moment while we fetch your results.
Product Title
Hamilton™ Septa, Long Life GC Autosampler > Diameter: 11mm