Learn More
Thermo Scientific™ Sensaphone™ Telephone Dialing Systems
Remotely monitor equipment from any standard touch-tone phone with this telephone dialing system will report the status of every monitored condition.
16460.18 NOK
Spezifikation
For Use With (Equipment) | Model 1535 Monitor/Alarm System |
---|---|
Monitoring Options | Alarm |
Benachrichtigungen:
Not approved for sales requiring CE mark.
Produktcode | Marke | Type | Electrical Requirements | Preis | Menge & VerfĂĽgbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Type | Electrical Requirements | Preis | Menge & VerfĂĽgbarkeit | |||||
12773838
|
Thermo Scientific™
400182 |
Four Channels | 220 V 50 Hz |
16460.18 NOK
Each |
Estimated Shipment: 27-08-2025
Log in to see stock. |
Sie haben bereits ein Konto? Melden Sie sich an um den Preis und die VerfĂĽgbarkeit zu sehen. Neu hier? Registrieren Sie sich noch heute bei uns! |
||||
Beschreibung
Call the lab or office from anywhere and Thermo Scientific™ Sensaphone™ Telephone Dialing Systems will state the status of every monitored condition. These systems interface with any standard touch-tone phone to notify of alarm conditions.
- Can be used with Model 1535 Monitor/Alarm System that features programmable audible/visual alarms and updates/displays equipment conditions on an LCD readout
- Battery backup is standard
- Battery life up to 15 hr. after AC power has failed; up to 18 months with AC power on
Requires:
Six D-size alkaline batteries
Spezifikation
Model 1535 Monitor/Alarm System | |
Alarm |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfĂĽllen.