Learn More
SemperIt™ Semperguard™ Nitrile Xpert Protective Gloves
Click to view available options
Medium (7-8)
Description
Protects against chemical splashing and in the processing of fatty foods. Provides excellent chemical resistance to skin irritants.
- Manufactured from high quality soft blue nitrile film
- Textured outer surface for better grip especially in wet conditions
- Ambidextrous fit with rolled cuff
Specifications
Specifications
Certifications/Compliance | CE Class I, EN 420, EN 374, EN 455, PPE Category III |
Color | Blue |
Disposable | Single-use |
Hand | Ambidextrous |
Latex Free | Yes |
Material | Nitrile |
Powdered | Powder Free |
Size | Medium, 7, 8 |
Thickness (Metric) | 0.20 mm |
Glove Cuff Style | Rolled |
Show More |
Product Content Correction
Product Title
SemperIt™ Semperguard™ Nitrile Xpert Protective Gloves >