Learn More
Kartell™ Self-Sprung Yellow Forceps
Use these forceps for a variety of laboratory applications. Kartell™ Self-Sprung Yellow Forceps are constructed of POM for excellent stability.
Brand:  Kartell™ 0050300
 View more versions of this product
Product Code. 10683472
  • 861.00 NOK / Pack of 5
Only null left
Estimated Shipment: 25-07-2025
Description
  • Autoclavable
  • Yelow
  • Self-spring with rounded ends
  • Constructed of POM
Specifications
Specifications
5/Pack
253 mm
Forceps
POM
Forceps
Yellow Forceps
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Kartell™ Self-Sprung Yellow Forceps > Length Metric: 253mm