Learn More
Keystone Cleanroom Products Self Seal Tyvek™ Steam Sterilization Pouches
BIOPHARMAGREENER_CHOICE
Self Seal Tyvek™ Steam Sterilization Pouches provide an excellent barrier from microbial contamination.
Brand:  Keystone Cleanroom Products BBMGTVKSS0306
 View more versions of this product
Product Code. 17727827
  • 14725.00 NOK / Pack of 1000
Only null left
Estimated Shipment: 28-07-2025
Description
  • Great choice for equipment parts that are heavy, have sharp edges, or packaging that requires surface sanitization post-sterilization
  • DuPont Tyvek™/Polyester-based flexible film is lint-free, moisture and puncture resistant, extremely tough, and designed to be autoclaved
  • The Tyvek™ side offers high breathability while providing an excellent barrier from microbial contamination
  • The clear film side offers excellent clarity along with strength and durability
  • These bags feature a medical grade adhesive strip for convenient self-sealing
  • Each pouch has a CLASS A process indicator, which changes from pink to dark brown when exposed to steam
  • When the peel-able seals are opened, virtually no airborne particulates are generated
  • Recommended for sterilization of pharmaceutical equipment
  • Manufactured and packaged in controlled cleanroom environment
  • Eliminates the need for the use and validation of a Heat Sealer
Specifications
Specifications
Autoclavable
Self-seal
Single-use
Tyvek
Yes
6 x 3 in.
15.2 cm
7.6 cm
Certificate of Conformance (COC)
White
Steam Sterilization
Non-sterile
1000/Pk.
6 in.
3 in.
Autoclave Bag
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Keystone Cleanroom Products Self Seal Tyvek™ Steam Sterilization Pouches > Dimensions (L x W): 6 x 3 in.