missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novex™ SelectFX™ Alexa Fluor™ 488 Endoplasmic Reticulum Labeling Kit, for fixed cells
SelectFX™ Alexa Fluor™ 488 Endoplasmic Reticulum Labeling Kit, for fixed cells
Brand: Novex™ S34200
This item is not returnable.
View return policy
Description
The SelectFX Alexa Fluor 488 Endoplasmic Reticulum Labeling Kit provides all the reagents needed to label the endoplasmic reticulum in fixed cells. To detect the ER, the kit uses a primary antibody directed against the ER-associated protein disulfide isomerase (PDI) and an Alexa Fluor 488 dye-labeled secondary antibody. The fluorescence can be observed using the same optical filters used for fluorescein. The kit also includes cell fixative and permeabilization reagents and protocols for mammalian cell preparation and staining.Kit Attributes:
•Applications: Validated application for SelectFX Alexa Fluor 488 Endoplasmic Reticulum Labeling Kit is endoplasmic reticulum staining.
•Format: Slide(s)
•Detection Method: Fluorescence microscopy
•Product Size: One kit
The network of folded membranes that comprises the endoplasmic reticulum (ER) in all eukaryotic cells constitutes greater than half of the total membrane in the average animal cell. The branching tubules and flattened sacs of the ER come in two forms: membrane bound by ribosomes (rough ER) or unbound by ribosomes (smooth ER). The ER has a central role in lipid and protein synthesis, protein chaperoning and folding, and calcium homeostasis. Abnormal protein folding in the ER is implicated in a number of diseases including cystic fibrosis and Alzheimer's disease.
The SelectFX Alexa Fluor 488 Endoplasmic Reticulum Labeling Kit provides all the reagents needed to label the ER in fixed cells. To detect the ER, the kit uses a primary antibody directed against the ER-associated protein disulfide isomerase (PDI) and an Alexa Fluor 488 dye-labeled secondary antibody; fluorescence is observed using standard fluorescein filters. The kit also includes cell fixative and permeabilization reagents and protocols for mammalian cell preparation and staining.

Specifications
Slide(s) | |
1 kit | |
Alexa Fluor™, SELECTFX™ | |
Alexa Fluor Dyes |
Fluorescence Microscope | |
Fluorescence | |
Room Temperature | |
Endoplasmic Reticulum Label |
SelectFX Alexa Fluor 488 Endoplasmic Reticulum Labeling Kit
Version: 05-31-2007
File size: 114 kB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novex™ SelectFX™ Alexa Fluor™ 488 Endoplasmic Reticulum Labeling Kit, for fixed cells > 1 kit
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction