Learn More
3M™ SecureFit™ SF600 Series Safety Glasses
Comprised of a semi-rimless lens and fixed-length temple arm. 3M™ SecureFit™ SF600 Series Safety Glasses include an integrated side shield for added protection.
37.70 NOK - 995.00 NOK
Specifications
Frame Color | Gray |
---|---|
Lens Material | Polycarbonate |
Frame Material | Polycarbonate |
UV Protection | Yes |
Product Code | Brand | Lens Coating | Lens Type | Lens Tint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Lens Coating | Lens Type | Lens Tint | Price | Quantity & Availability | |||||
15727727
|
3M™
7100112718 |
Scratch-resistant | I/O Mirror | N/A | |||||||
15747727
|
3M™
7100112721 |
Scratch-resistant | Welding shade 1.7 |
363.00 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15757727
|
3M™
7100112724 |
Scratch-resistant | Welding shade 3.0 |
363.00 NOK
Each |
Estimated Shipment: 21-07-2025
Log in to see stock. |
||||||
15797717
|
3M™
7100112715 |
Anti-fog, Scratch-resistant | Amber |
363.00 NOK
Each |
Estimated Shipment: 16-07-2025
Log in to see stock. |
||||||
15737727
|
3M™
7100112720 |
Scratch-resistant | Clear | N/A | |||||||
15787717
|
3M™
7100112713 |
Scratch-resistant | Polarized | Gray |
995.00 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
|||||
15707727
|
3M™
7100112716 |
Anti-fog, Scratch-resistant | Gray |
281.00 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15717727
|
3M™
7100112717 |
Anti-fog, Scratch-resistant | Clear |
246.00 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
15777717
|
3M™
7100112712 |
Scratch-resistant | Welding shade 5.0 |
270.00 NOK
Each |
Estimated Shipment: 01-07-2025
Log in to see stock. |
||||||
Description
- Optical class 1 lens suitable for prolonged use
- Design provides coverage and a good field of vision based on two lenses in a semi-rimless design
- Offers protection against Ultraviolet (UV) radiation in accordance with European standards
- Weight: 27g
- 3M™ Pressure Diffusion Temple Technology for secure and comfortable fit
- Soft nose pads for comfortable fit
- Superior Scotchgard™ anti-fog coating meeting K and N on the standard clear, gray, and amber lenses
Specifications
Gray | |
Polycarbonate | |
Polycarbonate | |
Yes |
Product Title
3M™ SecureFit™ SF600 Series Safety Glasses