Learn More
3M™ SecureFit™ SF600 Series Safety Glasses with Foam Insert
Description
- Optical class 1 lens suitable for prolonged use
- Design provides coverage and a good field of vision based on two lenses in a semi-rimless design
- Offers protection against Ultraviolet (UV) radiation in accordance with European standards
- Weight: 27g
- 3M™ Pressure Diffusion Temple Technology for secure and comfortable fit
- Soft nose pads for comfortable fit
- Superior Scotchguard anti-fog coating
- Includes Foam Insert
Specifications
Specifications
Lens Coating | Anti-fog |
Lens Tint | Gray |
Frame Color | Gray |
Includes | Foam insert |
Lens Material | Polycarbonate |
Frame Material | Polycarbonate |
UV Protection | Yes |
Videos
-
3M SecureFit protective eyewear
-
3M SecureFit protective eyewear 600 series
Product Content Correction
Product Title
3M™ SecureFit™ SF600 Series Safety Glasses with Foam Insert >