Learn More
3M™ Secure Click™ Quantitative Fit Test Adaptor Kit HF-800-03
Click to view available options
:
Head harness assembly
Description
- Use to conduct quantitative fit testing on 3M™ Secure Click™ Half Mask Reusable Respirators with speaking diaphragm
- Includes head harness assembly
Specifications
Specifications
| Product Type | Adapter Kit |
| For Use With (Equipment) | Conducting Quantitative Fit Testing on 3 M™ Secure Click™ Half Mask Reusable Respirators HF-800 SD Series |
| Includes | Head harness assembly |
Product Content Correction
Product Title
3M™ Secure Click™ Quantitative Fit Test Adaptor Kit HF-800-03 >