Learn More
Sartorius™ Secura™ Precision Laboratory Balance
Laboratory balance with internal calibration and continuous real-time level monitoring.
Brand: Sartorius™ SECURA3101-1S
This item is not returnable.
View return policy
Description
Advanced Level Monitoring and Security Secura™ Precision Balances comes with a host of advanced features to ensure the most accurate and precise results possible.
- Equipped with an advanced internal monitoring system that automatically calibrates your balance according to your password-protected menu settings
- Automatic calibration and adjustment system monitors and reacts to temperature changes, eliminating result errors and saving time
- Fully compliant data transfer capabilities and SQmin function to ensure reliable USP compliance
- Choose from three levels of security options to customize to the needs of your laboratory
- Ships with a series of integrated applications that significantly expand the potential of your balance from day one
- Included applications allow researchers to carry out advanced weight measurements of mixtures, measure the density of both liquid and solid samples, make complex multi-step conversions and create calibration and weight statistical measurement reports
- Balances arrive with an easy-to-navigate SmartTouch user display for easy menu navigation and application selection right out of the box
- One-Touch draft shield is designed to minimize ambient effects on measurements while protecting the integrity of your samples
Specifications
Council Directive 2009/23/EC, EN 45501:1992, OIML R76:2006 | |
Color Touchscreen | |
Stainless Steel | |
0.9 s |
3100 g | |
100 mg | |
Electronic Toploading Balances | |
100/240 V, 50/60 Hz |