Learn More
OHAUS™ Sectional Block
Hotplate-Stirrer accessory
Brand:  OHAUS™ 30500578
2760.91 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 16670462
  • 2835.00 NOK / Each
Only null left
Estimated Shipment: 23-07-2025
Description
Guardian accessories allow end users to have superb versatility. They can create Mini Lab Frames utalizing our large selection of accessories such as Rods, Clamps, Lab Lifts, Uni Blocks, Sectional Blocks, Splash Shields, Stir Bars, Temperature Probes, Achiever Overhead Stirrers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
16 mm Test Tubes
Sectional Block
Metal
Certificates
Promotions
Promotions
Product Content Correction
Product Title
OHAUS™ Sectional Block > 16 mm Test Tubes