Learn More
Welch™ Glass, Spare Flask
Capacity Metric: 2 L
Brand: Welch™ 112523-1
This item is not returnable.
View return policy
Description
Specifications
2 L | |
Glass | |
Aspirator Bottle |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Welch™ Glass, Spare Flask > Capacity: 2L