Learn More
SeaKem™ LE Agarose, Lonza™
Ideal for routine analysis of nucleic acids by gel electrophoresis and blotting. SeaKem™ LE Agarose sharply resolves DNA and provides consistent resolution from lot-to-lot.
Brand:  Lonza 50000
 View more versions of this product
Product Code. 11570717
  • 4855.00 NOK / Each
Estimated Shipment: 08-04-2025
to see stock.
Explore more special offers
Description
  • For DNA analysis/blotting
  • Molecular Biology Grade
  • EEO 0.09–0.13
  • Gelling temperature 36° ± 1.5°C
  • DNase/RNase-free
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SeaKem LE Agarose
125 g
Product Title
SeaKem™ LE Agarose, Lonza™ > 125g