Learn More
Cytiva SE260/SE250 MiniVertical Electrophoresis System Replacement Parts: Plates
Brand: Cytiva SE262N-5
This item is not returnable.
View return policy
Description
Specifications
HE260 Mini Vertical Unit | |
Alumina | |
1.05 cm |
Notched Alumina Plate | |
Rectangular | |
0.1 cm |
Product Content Correction
Product Title
Cytiva SE260/SE250 MiniVertical Electrophoresis System Replacement Parts: Plates > Plate; Notched Alumina