missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDHB Monoclonal Antibody (21A11AE7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 459230
This item is not returnable.
View return policy
Description
Positive control: Human heart mitochondria.
SDHB Monoclonal antibody specifically detects SDHB in Bovine, Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
SDHB | |
Monoclonal | |
1 mg/mL | |
HEPES buffered saline with 0.02% sodium azide | |
A5PL98, P21913, Q3T189, Q007T0, Q9CQA3, P21912 | |
SDHB | |
Bovine Heart Complex II, with traces of Complex III. | |
100 μg | |
Primary | |
Bovine, Human, Mouse, Rat | |
Antibody | |
IgG2a κ |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot | |
21A11AE7 | |
Unconjugated | |
SDHB | |
0710008N11Rik; CWS2; fj46d06; FLJ92337; I79_022285; Ip; iron-sulfur subunit; iron-sulfur subunit of complex II; OTTHUMP00000044624; PGL4; SDH; SDH1; SDH2; Sdhb; SDHB protein; SDHB; sdhB; SDHIP; succinate dehydrogenase [ubiquinone] iron-sulfur subunit; Succinate dehydrogenase [ubiquinone] iron-sulfur subunit, mitochondrial; succinate dehydrogenase complex iron sulfur subunit B; succinate dehydrogenase complex subunit B; succinate dehydrogenase complex subunit B, iron sulfur (Ip); succinate dehydrogenase complex, subunit B, iron sulfur (Ip); succinate dehydrogenase Ip subunit; succinate dehydrogenase iron-protein subunit; wu:fj46d06 | |
Mouse | |
IgG fraction | |
RUO | |
67680, 562149, 6390, 286840, 298596, 414412 | |
4° C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction