Learn More
Schneider Gerd™ Forceps
Use these forceps for a variety of applications. Schneider Gerd™ Forceps are made of stainless steel with blunt tips.
Brand:  Schneider Gerd™ 8111
Product Code. 10470724
  • 54.00 NOK / Each
Only null left
Estimated Shipment: 14-08-2025
Description
  • Stainless steel blunt-end forceps
  • For dissection
Specifications
Specifications
Blunt
115 mm
Blunt ended
Stainless Steel
Forceps
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Schneider Gerd™ Forceps > Stainless steel; Blunt ended; Length: 115mm