Learn More
SCH-1473759 hydrochloride, MedChemExpress
MedChemExpress SCH-1473759 hydrochloride is an aurora inhibitor with IC50s of 4 and 13 nM for aurora A and B, respectively.
Brand:  Medchem Express HY-10483
Encompass_Preferred
 View more versions of this product
Product Code. 16440116
  • 2354.75 NOK / 2mg
Description
SCH-1473759 hydrochloride is an aurora inhibitor with IC50s of 4 and 13 nM for aurora A and B, respectively.
Specifications
Specifications
SCH-1473759 hydrochloride
1094067-13-6
2 mg
C20H27ClN8OS
DMSO : 70 mg/mL (151.19 mM; Need ultrasonic)
463.0
98.04%
Solid
Off-White
Research
Cancer-Kinase/protease
[H]Cl.CC1=CN2C(C(NC3=CC(CN(C(C)(CO)C)CC)=NS3)=N1)=NC=C2C4=CNN=C4
463.0
Research
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
SCH-1473759 hydrochloride, MedChemExpress