Learn More
Sartorius™ Glass Fiber Sample Pads, for Infrared Moisture Analyzers
For use with all Denver Moisture Analyzers.
Brand: Sartorius™ 6906941
1333.65 NOK valid until 2025-07-11
Use promo code "25721" to get your promotional price.
This item is not returnable.
View return policy
Description
- Ideal for samples with a high moisture content
- Prevent crusting or splattering of samples while reducing analysis time
- 90mm dia.
Specifications
Denver IR-200 Infrared Moisture Analyzer | |
Glass Fiber | |
90 mm diameter | |
Sample Pad |
Product Content Correction
Product Title
Sartorius™ Glass Fiber Sample Pads, for Infrared Moisture Analyzers > 4.00 in. (120mm) dia.