Learn More
Sartorius Balance Density Determination Kits
GREENER_CHOICE
Use with Sartorius balances to determine the density of solids and liquids
Brand:  Sartorius YDK01MS
13336.61 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15229134
  • 13675.00 NOK / Each
Description
All kits include
  • 2 Beakers (7mm and 55mm dia.)
  • Bar frame
  • Thermometer with retainer clip
  • Glass plummet
  • Sieve for immersing samples (pan hanger assembly)
  • Sample holder (pan hanger assembly)
  • Metal platform
CPA Kit includes
  • Gasket for ME models
  • Compensating disk for ME235S/P, ME254S models
  • 3 adapters
Cubis Kits include
  • Adapter for LA/LP models with analytical draft shield
Extended ED Kit includes
  • Adapters for LA/LP models: 1 with and 1 without analytical draft shield
  • Compensating disk for LA/LP2300D models
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cubis Balance
2 Beakers (7 mm and 55 mm dia.), Bar Frame, Thermome
Density Determination Kit
Determining the density of solids and liquids
Cubis I
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius Balance Density Determination Kits > Cubis, less than 1mg