Learn More
SART3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18025-1-AP-150UL
 View more versions of this product
Product Code. 16848054
  • 4796.50 NOK / 150µL
Explore more special offers
Description
Description
The protein encoded by this gene is an RNA-binding nuclear protein that is a tumor-rejection antigen. This antigen possesses tumor epitopes capable of inducing HLA-A24-restricted and tumor-specific cytotoxic T lymphocytes in cancer patients and may be useful for specific immunotherapy. This gene product is found to be an important cellular factor for HIV-1 gene expression and viral replication. It also associates transiently with U6 and U4/U6 snRNPs during the recycling phase of the spliceosome cycle. This encoded protein is thought to be involved in the regulation of mRNA splicing.
Specifications
Specifications
SART3
Polyclonal
Unconjugated
SART3
DSAP1, hSART 3, KIAA0156, P100, p110, p110(nrb), RP11 13G14, SART 3, SART3, TIP110
Rabbit
Antigen Affinity Chromatography
RUO
304582, 53890, 9733
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q15020, Q9JLI8
Sart3
SART3 Fusion Protein Ag12590
150 μL
Primary
Human, Mouse, Rat