Learn More
Thermo Scientific Chemicals Sarcosine, 98%
Product Code. 10041860
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 107-97-1
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00004279
InChI Key FSYKKLYZXJSNPZ-UHFFFAOYSA-N
Synonym N-Methylglycine
PubChem CID 1088
ChEBI CHEBI:15611
IUPAC Name 2-(methylamino)acetic acid
SMILES CNCC(O)=O
Specifications
Color Colorless or White to Yellow
Physical Form Crystals or Powder
Quantity 500 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Assay Percent Range 98%
Linear Formula CH3NHCH2CO2H
Water 2% max.
Formula Weight 89.09
Percent Purity ≥97.5% (total base)
Chemical Name or Material Sarcosine
Show More Show Less
Product Identifier
  • Sarcosine