Learn More
Notrax Sani-Flex™ Specially Designed for the Lab Industry
Specially designed for the lab industry, Sani-Flex™ anti-fatigue matting provides the optimum combination of comfort, anti-slip surface, quality and durability, as well as oil/grease and chemical resistance. Its smooth surface and rounded off edges make it easy to clean.
832.00 NOK - 1515.00 NOK
Specifications
Color | Gray |
---|---|
Material | Nitrile Rubber |
Type | Industrial and Commercial Matting |
Product Code | Brand | Length (English) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (English) | Price | Quantity & Availability | ||||||
17792178
|
Notrax
526S1824GY |
2 ft. |
832.00 NOK
Each |
Estimated Shipment: 04-08-2025
Log in to see stock. |
||||||
17782178
|
Notrax
526S0023GY |
36 in. |
1515.00 NOK
Each |
Estimated Shipment: 30-07-2025
Log in to see stock. |
||||||
Description
- Specially designed for the lab industry, Sani-Flex™ anti-fatigue matting provides the optimum combination of comfort, anti-slip surface, quality and durability, as well as oil/grease and chemical resistance
- Its smooth surface and rounded off edges make it easy to clean
- Made of tough rubber compounds, designed to yield a long service life
- Aggressive slip-resistant surface
Specifications
Gray | |
Nitrile Rubber | |
Industrial and Commercial Matting |