Learn More
Rheodyne Sample loop (7125, 7010) SST 1/16 inch O.D.,0.020 inch (0.51mm) I.D. 50µL
Brand:  Rheodyne 02016-36
Sample loop (7125, 7010) SST 1/16 inch O.D.,0.020 inch (0.51mm) I.D. 50µL
Product Code. 13030193
  • 1190.00 NOK / Each
Product Title
Rheodyne Sample loop (7125, 7010) SST 1/16 inch O.D.,0.020 inch (0.51mm) I.D. 50µL >