Learn More
BRAND™ Sample Cups, PS, clear, for Technicon-Analyzer
Transparent polystyrene. For use with Technicon Analyzer.
Brand:  BRAND™ 115017
563.88 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10664501
  • 636.00 NOK / Pack of 1000
Estimated Shipment: 27-06-2025
Description
  • Sample vial
  • Polystyrene construction
  • Glass clear
  • IVD 4.0 mL
  • For use with Technicon Analyzer
TRUSTED_SUSTAINABILITY
Specifications
Specifications
17 mm
4 mL
1000
38 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BRAND™ Sample Cups, PS, clear, for Technicon-Analyzer > Capacity: 4mL