Learn More
BRAND™ Sample Cups, PS, clear, for Technicon-Analyzer
Transparent polystyrene. For use with Technicon Analyzer.
Brand:  BRAND™ 115015
398.34 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10165261
  • 446.00 NOK / Pack of 1000
Only null left
Estimated Shipment: 14-07-2025
Description
  • Sample vial
  • Polystyrene construction
  • Glass clear
  • IVD 1.5 mL
  • For use with Technicon Analyzer
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Transparent
Polystyrene
1000
22.7 mm
Technicon Analyzer
12.2 mm
1.5 mL
Polystyrene
1000 Pk.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BRAND™ Sample Cups, PS, clear, for Technicon-Analyzer > Capacity: 1.5mL