Learn More
BRAND™ Sample Cups, PS, clear, for Technicon-Analyzer
Transparent polystyrene. For use with Technicon Analyzer.
Brand: BRAND™ 115016
356.45 NOK valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Description
- Polystyrene
- Transparent
Specifications
Polystyrene | |
2.0 mL | |
1000 |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now