Learn More
S Murray and Co™ SAMCO™ Dressing Scissors
Use these scissors for a variety of applications. S Murray™ SAMCO™ Dressing Scissors comply with the requirements of the Medical Devices Directive 93/42/EEC, with full traceability maintained throughout the manufacturing process.
50.85 NOK - 64.90 NOK
Specifications
Product Type | Dressing Scissors |
---|---|
Blade Style | Straight |
Length (Metric) | 125 mm |
Material | Stainless Steel |
Product Code | Brand | Blade Style | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Blade Style | Price | Quantity & Availability | ||||||
12378099
|
S Murray and Co™
E812/01 |
Straight |
64.90 NOK
Each |
Estimated Shipment: 22-07-2025
Log in to see stock. |
||||||
12368099
|
S Murray and Co™
E801/02 |
Straight |
50.85 NOK
Each |
In Stock
Log in to see stock. ![]() |
||||||
Description
Use these scissors for a variety of applications. S Murray™ SAMCO™ Dressing Scissors comply with the requirements of the Medical Devices Directive 93/42/EEC, with full traceability maintained throughout the manufacturing process.
- Stainless Steel dressing scissors
Specifications
Dressing Scissors | |
Straight | |
125 mm | |
Stainless Steel |