missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals Salt Mixture No. 4164

Contains different variety of salts.

Brand:  MP Biomedicals 0290284950

 View more versions of this product

Product Code. 11485662

  • 31622.98 NOK / 50kg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Salt mixture containing calcium lactate and potassium phosphate.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Not Autoclavable
Single-use
Non-sterile
1 ft.
Animal Diet
50 kg
Room Temperature
0.3048 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.