missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint-Gobain Chemfluor™ Fluoropolymer Tubing
Ideal for semiconductor processing, biotechnology and pharmaceutical transfer, environmental sample preparation, chromatography, Type I Water, cryogenic, and hazardous chemicals
2500.00 NOK - 29584.00 NOK
Specifications
Certifications/Compliance | Complies with FDA criteria |
---|---|
For Use With (Application) | Ideal for semiconductor processing, biotechnology and pharmaceutical transfer, environmental sample preparation, chromatography, Type I Water, cryogenic, and hazardous chemicals |
Product Code | Brand | Diameter (English) Inner | Diameter (English) Outer | Material | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) Inner | Diameter (English) Outer | Material | Price | Quantity & Availability | |||
15639348
|
Saint-Gobain
TSFE14025006250 |
0.13 in. | 1/4 in. | Fluoropolymer |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11735179
|
Saint-Gobain
96000-02 |
0.063 in. | 0.13 in. | PTFE |
2500.00 NOK
Each |
Estimated Shipment: 04-10-2024
Log in to see stock. |
|||
Description
- Contains no additives or plasticizers
- Inert to virtually all chemicals
- Nonprotein binding
Order Info
In packs of 50 ft. (15m)
Compliance
Complies with FDA criteria
Specifications
Complies with FDA criteria | |
Ideal for semiconductor processing, biotechnology and pharmaceutical transfer, environmental sample preparation, chromatography, Type I Water, cryogenic, and hazardous chemicals |