Learn More
R&L Enterprises™ Bright Stainless Steel Rods
An excellent option for the construction of retort stands in the laboratory. R&L Enterprises™ Bright Stainless Steel Rods are durable and corrosion resistant.
94.00 NOK - 488.80 NOK
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Description | Price | Quantity & Availability | ||||
11795386
|
R&L Enterprises™
3108/33 |
750 mm | 12.5 mm | Threaded |
342.00 NOK
Each |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||
11785386
|
R&L Enterprises™
3108/13 |
500 mm | 10 mm | Threaded |
171.00 NOK
Each |
In Stock
Log in to see stock. ![]() |
||||
11759327
|
R&L Enterprises™
3109/11 |
500 mm | 12.5 mm | Plain (without thread) |
94.00 NOK
Each |
Estimated Shipment: 13-11-2025
Log in to see stock. |
||||
11705396
|
R&L Enterprises™
3108/43 |
1000 mm | 12.5 mm | Threaded |
488.80 NOK
Each |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||
12328149
|
R&L Enterprises™
3108/23 |
600 mm | 12.5 mm | Threaded |
206.00 NOK
Each |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||
Description
- Durable
- Easy-to-clean
- Corrosion resistant
- Stainless steel
Specifications
Stainless Steel | |
Rod |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
R&L Enterprises™ Bright Stainless Steel Rods