Learn More
SymLine Safety Waste Collector
SymLine is the modular disposal system for liquid waste. It connects several laboratory workstations, HPLC systems and fume cupboards to a central disposal or collection point, such as a safety cabinet. Made of electrostatic conductive PTFE connects capillaries and tubes of the HPLC system with the disposal system.
Brand: SymLine 306491-EL
884.85 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
SymLine Safety Waste Collectors route the waste tubes from your HPLC system directly into the disposal system. A choice of several connections offers you complete freedom. Fittings for different capillary diameters are included in the scope of delivery.
- Made of electrically conductive PTFE-EL material to prevent static discharge
- Available with different numbers of tube connections
- For use on a SymLine pipe system for liquid waste disposal
Specifications
Black | |
PTFE-EL | |
Safety Waste Collector |
3 x PFA-fittings for capillaries with (OD) 1.6/2.3/3.2 mm | |
GL45 |