Learn More
SymLine Safety Waste Collector
SymLine is the modular disposal system for liquid waste. It connects several laboratory workstations, HPLC systems and fume cupboards to a central disposal or collection point, such as a safety cabinet. Made of electrostatic conductive PTFE connects capillaries and tubes of the HPLC system with the disposal system.
Brand:  SymLine 306494-EL
1036.92 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 17815714
  • 1215.00 NOK / 1 set
Estimated Shipment: 20-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
SymLine Safety Waste Collectors route the waste tubes from your HPLC system directly into the disposal system. A choice of several connections offers you complete freedom. Fittings for different capillary diameters are included in the scope of delivery.
  • Made of electrically conductive PTFE-EL material to prevent static discharge
  • Available with different numbers of tube connections
  • For use on a SymLine pipe system for liquid waste disposal
Specifications
Specifications
Black
PTFE-EL
Safety Waste Collector
5 x PFA-fittings for capillaries with (OD) 1.6/2.3/3.2 mm
GL45